90129
1-Dodecanethiol
analytical standard
Synonym(s):
n-Dodecyl mercaptan, Dodecyl mercaptan, Lauryl mercaptan, Mercaptan C12, NDM
About This Item
Recommended Products
grade
analytical standard
vapor density
7 (vs air)
assay
≥98.0% (GC)
form
liquid
shelf life
limited shelf life, expiry date on the label
refractive index
n20/D 1.457-1.461
n20/D 1.459 (lit.)
bp
266-283 °C (lit.)
density
0.845 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCCCCS
InChI
1S/C12H26S/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3
InChI key
WNAHIZMDSQCWRP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Recommended products
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1A
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
262.4 °F - closed cup
flash_point_c
128 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service