Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90051

Supelco

2,4,6-Triallyloxy-1,3,5-triazine

analytical standard

Synonym(s):

Triallyl cyanurate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O3
CAS Number:
Molecular Weight:
249.27
Beilstein/REAXYS Number:
235560
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

assay

≥96.5% (GC)

form

solid

shelf life

limited shelf life, expiry date on the label

bp

156 °C

mp

26-27 °C

density

1.11 g/mL at 30 °C

SMILES string

C=CCOc1nc(OCC=C)nc(OCC=C)n1

InChI

1S/C12H15N3O3/c1-4-7-16-10-13-11(17-8-5-2)15-12(14-10)18-9-6-3/h4-6H,1-3,7-9H2

InChI key

BJELTSYBAHKXRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

347.9 °F

flash_point_c

175.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service