89991
1-Octanethiol
analytical standard
Synonym(s):
n-Octyl mercaptan, Mercaptan C8, Octyl mercaptan
About This Item
grade
analytical standard
assay
≥98.5% (GC)
form
liquid
shelf life
limited shelf life, expiry date on the label
refractive index
n20/D 1.452 (lit.)
bp
197-200 °C (lit.)
density
0.843 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCS
InChI
1S/C8H18S/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3
InChI key
KZCOBXFFBQJQHH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
154.4 °F - closed cup
flash_point_c
68 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service