Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89829

Supelco

Tetraethyl orthosilicate

analytical standard

Synonym(s):

Orthosilicic acid tetraethyl ester, TEOS, Tetraethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si(OC2H5)4
CAS Number:
Molecular Weight:
208.33
Beilstein/REAXYS Number:
1422225
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

vapor density

7.2 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

assay

≥99.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.382 (lit.)
n20/D 1.382-1.384

bp

168 °C (lit.)

density

0.933 g/mL at 20 °C (lit.)

SMILES string

CCO[Si](OCC)(OCC)OCC

InChI

1S/C8H20O4Si/c1-5-9-13(10-6-2,11-7-3)12-8-4/h5-8H2,1-4H3

InChI key

BOTDANWDWHJENH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Commonly used as a precursor to prepare xerogel
Will interact with dodecylamine in the formation of intercalation compounds of H+-magadiite and used in a study of mixed-metal bioactive glasses.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

113.0 °F - closed cup

flash_point_c

45 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service