861424-U
Total Petroleum Hydrocarbons (TPH) Mixture 1
certified reference material, 2000 μg/mL each component in dichloromethane: hexane (1:1)
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
product line
TraceCERT®
CofA
current certificate can be downloaded
feature
standard type calibration
packaging
ampule of 1 mL
concentration
2000 μg/mL each component in dichloromethane: hexane (1:1)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
petroleum
format
multi-component solution
storage temp.
2-30°C
Application
Other Notes
Legal Information
Analyte
signalword
Danger
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3
target_organs
Central nervous system, Nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
-5.8 °F
flash_point_c
-21 °C
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service