Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

861394-U

Supelco

Total Petroleum Hydrocarbons (TPH) Mixture 3

1000 μg/mL each component in carbon disulfide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in carbon disulfide

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Decane
    Dodecane
    Dotriacontane
    Eicosane
    Heptane
    Hexadecane
    Hexane
    Hexatriacontane
    Nonane
    Octacosane
    Octadecane
    Octane
    Tetracontane
    Tetracosane
    Tetradecane
    Tetratetracontane
    Undecane
    See All (17)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

target_organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-22.0 °F - closed cup

flash_point_c

-30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service