Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

861377

Supelco

EPA 8270 Base Nuetral Surrogate Mix 1

5000 μg/mL each component in methylene chloride, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
Product 861377 is not currently sold in your country. Contact Technical Service

grade

analytical standard

agency

EPA 8270

packaging

ampule of 5 × 5 mL

concentration

5000 μg/mL each component in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

storage temp.

room temp

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    2-Fluorobiphenyl
    Nitrobenzene-d5
    p-Terphenyl-d14

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service