Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

861299

Supelco

EPA 8260 Internal Standards Mix

2500 μg/mL each component in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502

grade

analytical standard

agency

EPA 8260

CofA

current certificate can be downloaded

packaging

ampule of 5 × 1 mL

concentration

2500 μg/mL each component in methanol

application(s)

environmental

format

multi-component solution

Storage temp.

2-8°C

Analyte

Description

    Chlorobenzene-d5
    1,4-Dichlorobenzene-d4
    1,4-Difluorobenzene
    Pentafluorobenzene

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service