Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

861176

Supelco

Aramite

certified reference material, 2000 μg/mL in methylene chloride, ampule of 1 mL

Synonym(s):

Aramite solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23ClO4S
CAS Number:
Molecular Weight:
334.86
UNSPSC Code:
12352200

grade

certified reference material
TraceCERT®

agency

EPA 8270

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C15H23ClO4S/c1-12(20-21(17)19-10-9-16)11-18-14-7-5-13(6-8-14)15(2,3)4/h5-8,12H,9-11H2,1-4H3

InChI key

YKFRAOGHWKADFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aramite is a carcinogenic acaricide,[1] widely used against six spotted mite, European red mite and others affecting crops.[2]

Application

Aramite may be used as an analytical reference standard for the quantification of the analyte in agricultural commodities using gas chromatography coupled to mass spectrometry and liquid chromatography with tandem mass spectrometry.[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mites Injurious to Economic Plants (1975)
Insecticides: Analytical Methods for Pesticides, Plant Growth Regulators, and Food Additives, Volume 2, 2(6), 1366-1408 (2013)
Mites Injurious to Economic Plants (1975)
P Carricaburu et al.
Toxicological European research. Recherche europeenne en toxicologie, 2(4), 195-198 (1979-07-01)
Aramite was injected to white mice and the electroretinograms recorded in response to electronic flashes. The following phenomena were remarked: a more and more important flattening of a-waves; a simultaneous collapse of b-waves; a more and more delayed return of
F Moriya et al.
Nihon hoigaku zasshi = The Japanese journal of legal medicine, 45(2), 158-165 (1991-04-01)
Forensic toxicological examination was performed on a person who was suspected of poisoning by Azomite emulsion (an acaricide), Roundup (a herbicide) and/or unidentified agricultural chemical and died after 4 days. In Japan, production of Azomite emulsion has been stopped since

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service