843280
TSKgel® IEXPAK PW Bulk Media Sampler
phase various, DEAE-5PW, SP-5PW, SuperQ-5PW, bottle of 3 × 25 mL, 30 μm particle size
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115700
product line
TSKgel®
form
slurry
packaging
bottle of 3 × 25 mL
manufacturer/tradename
Tosoh 43280
parameter
20 bar max. pressure
technique(s)
LPLC: suitable
matrix
hydroxylated methacrylic polymer
particle size
30 μm
operating pH
2-13
separation technique
anion exchange
Looking for similar products? Visit Product Comparison Guide
General description
LabPak products are multi-milliliter containers of TOYOPEARL and TSKgel bulk media products. Typically they contain 3 or 4 different ligand types offered for a particular chromatography mode. They are useful for developmental engineers who wish to familiarize themselves with resin physical properties in different buffer systems. The larger resin amounts in LabPak products allow the packing of wider ID and longer columns than available in the ToyoScreen products. This helps the developmental chemist or engineer to better measure under actual packing conditions such properties as dynamic binding capacity, selectivity and column efficiency.
Specifications
Clean in place with 0.5 M NaOH or 0.1 M HCl.
Physical form
72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.
Legal Information
TSKgel is a registered trademark of Tosoh Corporation
related product
signalword
Danger
hcodes
Hazard Classifications
Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service