Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

843176

TSKgel® Ether-5PW (30) Bulk Media

bottle of 25 mL, phase ether, 30 μm particle size

Synonym(s):

TSKgel® Ether-5PW HIC Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product line

TSKgel®

form

slurry

packaging

bottle of 25 mL

manufacturer/tradename

Tosoh 43176

parameter

20 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

ether phase

particle size

30 μm

operating pH

1-13

capacity

20 g/L±10 adsorption capacity (lysozyme)

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel Ether-5PW resin is the highest hydrophilicity of the five HIC ligands offered by Tosoh Bioscience. It is useful for the separation of very hydrophobic molecules such as membrane proteins.  It is typically used for high performance chromatography and polishing steps. It has the same selectivity as the larger particle Toyopearl Ether-650 products. It often is used instead of cross-linked dextrans with exclusion limits of 10,000 MW.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

103.1 °F

flash_point_c

39.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service