Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

843125

TOYOPEARL® HICPAK-C Bulk Media Sampler

phase Phenyl, Butyl, Hexyl-650C, pkg of 3 × 25 mL, 100 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
Product 843125 is not currently sold in your country. Contact Technical Service

product line

TOYOPEARL®

form

slurry

packaging

pkg of 3 × 25 mL

manufacturer/tradename

Tosoh 43125

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

Phenyl, Butyl, Hexyl-650C phase

particle size

100 μm

operating pH

1-13

separation technique

hydrophobic interaction (HIC)

General description

LabPak products are multi-milliliter containers of TOYOPEARL and TSKgel bulk media products. Typically they contain 3 or 4 different ligand types offered for a particular chromatography mode. They are useful for developmental engineers who wish to familiarize themselves with resin physical properties in different buffer systems. The larger resin amounts in LabPak products allow the packing of wider ID and longer columns than available in the ToyoScreen products. This helps the developmental chemist or engineer to better measure under actual packing conditions such properties as dynamic binding capacity, selectivity and column efficiency.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service