Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

822891

TSKgel® GMHHR-H (20 µm and 30 µm) HT2 HPLC Columns

L x I.D. 7.5 cm × 7.5 mm, polystyrene-divinylbenzene phase

Synonym(s):

TSKgel® GPC Columns for Applications up to 220° C

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$957.00

$957.00


Check Cart for Availability


Select a Size

Change View
1 EA
$957.00

About This Item

UNSPSC Code:
41115711

$957.00


Check Cart for Availability

Product Name

TSKgel® GMHHR-H (20) and (30) HT2 Guard Column, phase polystyrene-divinylbenzene, L × I.D. 7.5 cm × 7.5 mm, 30 μm particle size

material

stainless steel

agency

suitable for USP L21

product line

TSKgel®

manufacturer/tradename

Tosoh 22891

parameter

1 mL/min flow rate
220 °C temperature

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 7.5 mm

matrix

polystyrene-divinylbenzene

matrix active group

polystyrene-divinylbenzene phase

particle size

30 μm

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel Ultra-High Temperature columns are for the analysis of organic-soluble polymers at extremely elevated temperatures. These columns are for applications up to 220°C and are shipped in o-dichlorobenzene (ODCB), which can be exchanged for 1-chloronaphthalene and trichlorobenzene.
The TSKgel mixed bed columns are packed with particles of different pore sizes that are blended to provide an extended linear calibration curve. Mixed bed columns are used when a broad range of polymer molar masses is expected in the sample.

Physical form

ODCB can be exchanged by 1-chloronaphthalene and trichlorobenzene.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service