Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

821999

TSKgel® NH2-100 DC Direct Connect (3 µm) HPLC Columns

L x I.D. 5 cm × 4.6 mm, amine phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

Product Name

TSKgel® NH2-100 DC Direct Connect HPLC Column, phase amine, L × I.D. 5 cm × 4.6 mm, 3 μm particle size

material

stainless steel

agency

suitable for USP L8

product line

TSKgel®

manufacturer/tradename

Tosoh 21999

parameter

0-100% organic solvent
1.0 mL/min standard flow rate
10-50 °C temperature
50 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

5 cm × 4.6 mm

surface area

450 m2/g

matrix

spherical silica

matrix active group

amine phase

particle size

3 μm

operating pH

2.0-7.5

separation technique

anion exchange
hydrophilic interaction (HILIC)
normal phase

General description

TSKgel NH2-100 DC column connects directly to TSKgel reversed phase columns. The DC in TSKgel NH2-100 DC emphasizes this Direct Connect aspect. This column has the same packing material as the TSKgel NH2-100 columns, so therefore shows high retention for hydrophilic compounds/ions. A male-type outlet endfitting enables the direct connection to the screw-type endfitting of a TSKgel reversed phase column. This allows for the simultaneous (linear) gradient separation of an active pharmaceutical ingredient (API) and its counterion without the loss of column efficiency normally experienced when connecting two columns with capillary tubing.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service