Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

821985

TOYOPEARL® Q-600C AR Bulk Media

bottle of 100 mL, phase ammonium ion, quaternary, 100 μm particle size

Synonym(s):

TOYOPEARL® Q-600C AR Anion Exchange Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
Product 821985 is not currently sold in your country. Contact Technical Service

product line

TOYOPEARL®

feature

counter ion Cl-

packaging

bottle of 100 mL

manufacturer/tradename

Tosoh 21985

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

ammonium ion, quaternary phase

particle size

100 μm

operating pH

2-13

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl Q-600C AR resin is a strong anion exchanger designed to be more alkaline resistant (AR) than Toyopearl QAE-550C resin. It has a slightly larger pore diameter than Toyopearl QAE-550C resin, but its ligand attachment chemistry creates significantly more dynamic binding capacity (up to 3X higher) than Toyopearl QAE-550C resin. It tolerates caustic cleaning solutions up to 1.0 mol/L NaOH well with essentially no change in DBC. Its elution kinetics are good and its retentivity is slightly stronger than Toyopearl QAE-550C resin.

Specifications

Clean in place with 0.1 M HCl or 0.1 M NaOH (can tolerate exposure up to 1.0 M NaOH for several months).

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

104.0 °F - closed cup

flash_point_c

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service