Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

821963

TSKgel® SP-STAT (10 µm) HPLC Columns

L x I.D. 3.5 cm × 3 mm, sulfonic acid phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,690.00

$1,690.00


Available to ship onApril 29, 2025Details



Select a Size

Change View
1 EA
$1,690.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

$1,690.00


Available to ship onApril 29, 2025Details


Product Name

TSKgel® SP-STAT HPLC Column, phase sulfonic acid, L × I.D. 3.5 cm × 3 mm, 10 μm particle size

material

stainless steel hardware

product line

TSKgel®

feature

counter ion Na+

manufacturer/tradename

Tosoh SP-STAT, 21963

parameter

no limitation salt concentration
≤50% organic solvent
1.0-2.0 mL/min standard flow rate
10-60 °C temperature
100 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

3.5 cm × 3 mm

matrix

polymer particle platform

matrix active group

sulfonic acid phase

particle size

10 μm

operating pH

3-10

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel SP-STAT columns are packed with 7 and 10μm hydrophilic non-porous resin particles of which the surface consists of an open access network of multi-layered cation exchange groups. The innovative bonding chemistry, combined with a relatively large particle size, results in a respectable loading capacity and a low operating pressure. Applications for the TSKgel SP-STAT columns include the separation of proteins, protein aggregates, charge isomers of monoclonal antibodies, PEGylated proteins, and peptide digests.

Physical form

Shipped in DI water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service