Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

821946

TOYOPEARL GigaCap® CM-650M Bulk Media

phase carboxylmethyl, bottle of 100 mL, 75 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
Product 821946 is not currently sold in your country. Contact Technical Service

product line

TOYOPEARL®

form

slurry

feature

counter ion Na+

packaging

bottle of 100 mL

manufacturer/tradename

Tosoh 21946

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

carboxylmethyl phase

particle size

75 μm

operating pH

3-13

capacity

>110 g/L adsorption capacity (human IgG)

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl GigaCap CM-650 resin is a high capacity cation exchange resin* optimized specifically for high throughput purification of IgG. Toyopearl HW-65 resin is the polymeric base bead, which is chemically modified to provide a high level of cationic binding sites for IgG. It exhibits typical dynamic binding capacities in the vicinity of 100 mg/mL for IgG. This new polymer product is stable at high linear velocities, has good pressure flow characteristics and will withstand back pressures up to 3 bar. The new higher capacity and back pressure stability of Toyopearl GigaCap CM-650 resin creates opportunities for increased throughput in downstream cation exchange purification steps.
Toyopearl GigaCap CM-650 resin has a different selectivity than Toyopearl GigaCap S-650 resin. With its unusually high capacity for a weak cationic exchange resin this selectivity may be exploited for either mAb or traditional protein purifications. Toyopearl GigaCap CM-650 resin also has excellent base stability in 0.5mol/L NaOH.
*specifically designed for packed bed use

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation
Toyopearl GigaCap is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service