Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

821928

TSKgel® ODS-140HTP (2.3 µm) HPLC Columns

L x I.D. 10 cm × 2.1 mm, octadecyl silane, multilayer phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

Product Name

TSKgel® ODS-140HTP HPLC Column, phase octadecyl silane, multilayer, L × I.D. 10 cm × 2.1 mm, 2.3 μm particle size

material

stainless steel

agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh 21928

extent of labeling

8% Carbon, endcapped

parameter

0-100% organic solvent
10-50 °C temperature
600 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

10 cm × 2.1 mm

surface area

100 m2/g

matrix

spherical silica

matrix active group

octadecyl silane, multilayer phase

particle size

2.3 μm

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel ODS-140HTP columns provide high resolution and short analyses time at moderate pressures, enabling high throughput separations. The polylayer bonding chemistry of the 2.3 μm particle size of these columns results in highly efficient and durable columns. The lower pressure drop reduces the burden on the hardware, allowing the TSKgel ODS-140HTP columns to be used with either UHPLC (up to 9000psi) or conventional HPLC systems.
For use in high throughput applications, including drug discovery, pharmacokinetics and peptide digest separations, TSKgel ODS-140HTP columns offer excellent peak shape for basic compounds.

Physical form

Shipped in acetonitrile.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service