Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

821888

TOYOPEARL® Phenyl-600 HIC Bulk Media

bottle of 100 mL, phase phenyl, 65 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
Product 821888 is not currently sold in your country. Contact Technical Service

product line

TOYOPEARL®

form

slurry

packaging

bottle of 100 mL

manufacturer/tradename

Tosoh 21888

parameter

≤50% organic solvent
3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

phenyl phase

particle size

65 μm

operating pH

1-13

capacity

55 g/L±10 adsorption capacity (lysozyme)

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl Phenyl-600 resin has the middle hydrophobicity of the five HIC ligands offered by Tosoh Bioscience. Its pore size has been optimized for the chromatography of monoclonal antibodies and it is typically used in an intermediate or polishing step. It is a very high recovery and high dynamic capacity resin for both large and small proteins. When used as the polishing step up to an extra 5% yield may be seen compared to other Phenyl resins. To see other products optimized for HIC purification of mAbs please see Toyopearl PPG-600 and Toyopearl Butyl-600 resins.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service