Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

821378

ToyoScreen® Hexyl-650C Process Development Column

phase C6 (hexyl), pkg of 6 × 1 mL, 100 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

material

aluminum seal
polypropylene and polyethylene frit

product line

ToyoScreen®

packaging

pkg of 6 × 1 mL

manufacturer/tradename

Tosoh 21378

parameter

≤50% organic solvent
0.2-1 mL/min flow rate
3 bar max. pressure
4 mL/min max. flow rate

technique(s)

HPLC: suitable

L × I.D.

3 cm × 6.4 mm

matrix

hydroxylated methacrylic polymer

matrix active group

C6 (hexyl) phase

particle size

100 μm

operating pH

1-13

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Every large scale commercial purification begins at a small scale in a method development lab by evaluating retention and selectivity of the target protein versus impurities, host cell proteins, DNA, etc., on multiple resins, often from different vendors. Depending on the constraints, the process development engineer can either purchase bulk resins to pack several small columns with different resins or take advantage of the speed and convenience of pre-packed columns for resin evaluation. Pre-packed ToyoScreen® Process Development Columns are available in 1 mL and 5 mL volumes packed with the most popular Toyopearl® resins. ToyoScreen columns can be used to quickly establish optimum elution conditions. Varying pH, salt type, gradient conditions and flow rate are common experimental parameters explored. Initial results from resin screening and optimization with ToyoScreen columns accurately predict the separation behavior at larger scale.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ToyoScreen is a registered trademark of Tosoh Corporation
Toyopearl is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service