Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

819690

TOYOPEARL® AF-Blue HC-650M Bulk Media

phase Cibacron Blue 3GA, 65 μm particle size, bottle of 1 L

Synonym(s):

TOYOPEARL® AF-Blue HC-650M Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
Product 819690 is not currently sold in your country. Contact Technical Service

description

Adsorption Capaacity (> 18 mg/mL (human serum albumin))
Shipping solvent (20% (v/v) ethanol)
phase filter (Blue)

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 19690

extent of labeling

>16 μmol/mL ligand density

parameter

pressure (max. 3 bar)

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

matrix active group

Cibacron Blue 3GA phase

particle size

65 μm

pore size

1000 Å

pH

4-9

capacity

>18 g/L adsorption capacity (human serum albumin)

separation technique

affinity

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl AF-Blue HC-650 resin is functionalized with Cibachron Blue F3G-A. It has high dynamic binding capacity for proteins, particularly albumin, and has improved caustic stability and lower dye leakage than traditional agarose materials

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service