Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

819013

TSKgel® CN-80Ts (5 µm) HPLC Columns

L x I.D. 1.5 cm × 3.2 mm, cyano phase

Synonym(s):

TSKgel® CN-80Ts Columns

Sign Into View Organizational & Contract Pricing

Select a Size

3 EA
$396.00

$396.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
3 EA
$396.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

$396.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

Product Name

TSKgel® CN-80Ts Guard Cartridge, phase cyano, L × I.D. 1.5 cm × 3.2 mm, 5 μm particle size, pkg of 3 ea

material

stainless steel

agency

suitable for USP L10

product line

TSKgel®

packaging

pkg of 3 ea

manufacturer/tradename

Tosoh CN-80Ts, 19013

extent of labeling

8% Carbon, monolayer, endcapped

parameter

0-100% organic solvent
10-50 °C temperature

technique(s)

HPLC: suitable

L × I.D.

1.5 cm × 3.2 mm

surface area

425 m2/g

matrix

silica particle platform

matrix active group

cyano phase

particle size

5 μm

pore size

80 Å

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Guard Cartridge for 4.6mmID TSKgel CN-80Ts columns, 3pk

Physical form

Shipped in 60% acetonitrile - 40% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

required but not provided

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service