Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

818768

TSKgel® ODS-80Ts QA HPLC Column

phase C18 (octadecyl), L × I.D. 15 cm × 2 mm, 5 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

material

stainless steel

agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh 18768

extent of labeling

15% Carbon, monolayer, endcapped

parameter

0-100% organic solvent
0.22 mL/min flow rate
10-50 °C temperature
200 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 2 mm

matrix

spherical silica

matrix active group

C18 (octadecyl) phase

particle size

5 μm

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel® ODS-80Ts QA columns were developed specifically for use by QA/QC departments that require highly reproducible separations. These columns are prepared from the same endcapped C18 packing material as TSKgel ODS-80Ts columns, but with narrower manufacturing specifications to meet the demand for high reproducibility.The variation between different lots of TSKgel ODS-80Ts QA packing material is minimized by selecting batches of TSKgel ODS-80Ts that fall within a very narrow range of specifications. In addition, each column must pass demanding specifications for efficiency (N) and peak asymmetry, as are spelled out in the Operating Conditions and Specifications sheet. The end result is TSKgel ODS-80Ts QA columns that exhibit an unparalleled level of reproducibility for retention, selectivity (k´), efficiency (N), and peak symmetry.

Physical form

Shipped in 70% methanol - 30% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service