Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

818557

TSKgel® Lipopropak XL (10 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, hydroxylated methacrylate phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

Product Name

TSKgel® Lipopropak XL HPLC Column, phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 10 μm particle size

material

stainless steel

product line

TSKgel®

manufacturer/tradename

Tosoh 18557

parameter

≤0.5 M salt concentration
≤20% organic solvent
1 mL/min flow rate
20 bar max. pressure
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

hydroxylated methacrylic polymer

matrix active group

hydroxylated methacrylate phase

particle size

10 μm

operating pH

2-12

separation technique

size exclusion (SEC)

General description

The TSKgel® Lipopropak XL column is a specialty column for the analysis of lipoprotein classes in serum using a dedicated eluent. The particles in TSKgel Lipopropak XL columns are spherical 10 micron porous polymethacrylate beads while the separation of the lipoproteins takes place by size exclusion. The TSKgel Lipopropak guard columns is packed with 17 micron polymethacrylate beads.

Physical form

Shipped in TSKeluent LP-1 (aqueous). Replace with appropriate mobile phase.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service