Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

818275

TSKgel® Super-Octyl (2.3 µm) HPLC Columns

L x I.D. 5 cm × 4.6 mm, C8 (octyl) phase

Synonym(s):

TSKgel® Reversed Phase HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

Product Name

TSKgel® Super-Octyl HPLC Column, phase C8 (octyl), L × I.D. 5 cm × 4.6 mm, 2.3 μm particle size

material

stainless steel hardware

agency

suitable for USP L7

product line

TSKgel®

manufacturer/tradename

Tosoh Super-Octyl, 18275

extent of labeling

5% Carbon, multilayer, endcapped

parameter

no limitation salt concentration
0-100% organic solvent
10-50 °C temperature
300 bar max. pressure
4 mL/min flow rate

technique(s)

HPLC: suitable

L × I.D.

5 cm × 4.6 mm

surface area

100 m2/g

matrix

silica particle platform

matrix active group

C8 (octyl) phase

particle size

2.3 μm

pore size

140 Å

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Physical form

Shipped in 70% methanol - 30% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service