Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

816651

TSKgel® ODS-80Tm (5 µm) HPLC Columns

L x I.D. 7.5 cm × 4.6 mm, C18 (octadecyl) phase

Synonym(s):

TSKgel® Reversed Phase HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

Product Name

TSKgel® ODS-80Tm HPLC Column, phase C18 (octadecyl), L × I.D. 7.5 cm × 4.6 mm, 5 μm particle size

material

stainless steel

agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh ODS-80Tm, 16651

extent of labeling

15% Carbon, monolayer, endcapped

parameter

0-100% organic solvent
0-45 °C temperature
1.2 mL/min flow rate
10 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 4.6 mm

surface area

425 m2/g

matrix

silica particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å pore diameter

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel ODS-80Tm is a general purpose column for the analysis of low MW pharmaceuticals, bases, nucleosides and nucleotides. Common applications include purity checks and peptide mapping. Bonded phase pore size is indicated by the number in the product description, in this case TSKgel ODS-80Tm has 80Å nominal pore size. The pore size of the base silica is 100Å.

Physical form

Shipped in 70% methanol - 30% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service