Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

814455

TSKgel® Tresyl-5PW (10 µm) HPLC Columns

L x I.D. 4 cm × 6 mm, tresyl phase

Synonym(s):

TSKgel® Tresyl-5PW HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

Product Name

TSKgel® Tresyl-5PW HPLC Column, phase tresyl, L × I.D. 4 cm × 6 mm, 10 μm particle size

material

stainless steel hardware

product line

TSKgel®

analyte chemical class(es)

proteins

manufacturer/tradename

Tosoh Tresyl-5PW, 14455

parameter

≤20% organic solvent
≤3 M salt concentration
>0 °C temperature
1.0 mL/min flow rate
10 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

4 cm × 6 mm

matrix

polymer particle platform

matrix active group

tresyl phase

particle size

10 μm

operating pH

2-12

capacity

>60 mg/g, dry resin coupling capacity (soybean trypsin inhibitor)

separation technique

affinity

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Unlike other TSKgel affinity columns, TSKgel Tresyl-5PW, which is derivatized with the 2,2,2-trifluroethanesulfonyl ligand, requires activation with a user-selected ligand containing amino, thiol, phenol, or imidazole groups. The resulting structure is literally a "custom" affinity ligand with excellent pH stability and minimal ligand loss due to leaching. TSKgel Tresyl-5PW readily reacts with amino or thiol groups to form stable covalent alkylamines or thioethers.

Applications include: glycoproteins, antigens, etc.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service