Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

814443

TSKgel® Heparin-5PW HPLC Column

phase sulfated glycosaminoglycan, L × I.D. 5 cm × 5 mm, 10 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

material

glass

product line

TSKgel®

analyte chemical class(es)

proteins

manufacturer/tradename

Tosoh 14443

parameter

≤20% organic solvent
≤3 M salt concentration
1.0 mL/min flow rate
20 bar max. pressure
4-30 °C temperature

technique(s)

HPLC: suitable

L × I.D.

5 cm × 5 mm

matrix

hydroxylated methacrylic polymer

matrix active group

sulfated glycosaminoglycan phase

particle size

10 μm

operating pH

5.5-9.0

capacity

>1.8 mg/mL, resin adsorption capacity (antithrombin III)

separation technique

affinity

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

High efficiency, resin-based TSKgel affinity columns (10 μm particles) separate or purify many enzymes and other proteins.

The heparin ligand in TSKgel Heparin-5PW affinity columns can interact with sample molecules by cation exchange as well as affinity chromatography. Cation exchange interaction takes place through O-sulfate and N-sulfate groups. The elution strength is similar to TSKgel SP-5PW.

In vivo, heparin prevents blood clotting as it reacts with clotting agents. This tendency is taken advantage of by using heparin to separate blood clotting factors such as antithrombin III, Factor VII, Factor IX,etc., and anti-heparin proteins such as glycoproteins, endoglycosidase, hyaluronidase, lipases, growth factors, RNA polymerases, and other nucleic acid-binding proteins.

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service