Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

813181

TSKgel® Aminopak (5 µm) HPLC Columns

L x I.D. 12 cm × 4.6 mm, sulphonic acid phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

Product Name

TSKgel® Aminopak HPLC Column, phase sulphonic acid, L × I.D. 12 cm × 4.6 mm, 5 μm particle size

material

stainless steel

product line

TSKgel®

manufacturer/tradename

Tosoh 13181

parameter

≤20% organic solvent
0.4 mL/min flow rate
25-80 °C temperature
80 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

12 cm × 4.6 mm

matrix

polystyrene-divinylbenzene

matrix active group

sulphonic acid phase

particle size

5 μm

operating pH

2-14

separation technique

cation exchange

Specifications

Clean in place with 0.2 M NaOH.
TSKgel® Aminopak is a specialty column for the analysis of free amino acids by ion exchange. The column is packed with polystyrene-based particles that have been functionalized with sulfonic acid groups. Detection is performed by post-column reaction with ninhydrin or o-phthalaldehyde. The primary application of TSKgel Aminopak is for the analysis of amino acids in food. No guard column products are offered to protect TSKgel Aminopak columns. It is recommended to use a frit or screen fiter (<2 micron porosity) between the injector and the column. Samples and mobile phases are best filtered and degassed through a 0.45 micron membrane filter.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service