Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808646

TSKgel® Chelate-5PW (13 µm) HPLC Columns

L x I.D. 15 cm × 21.5 mm, iminodiacetic acid phase

Synonym(s):

TSKgel® Chelate-5PW HPLC Column and Guardgel Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

Product Name

TSKgel® Chelate-5PW HPLC Column, phase iminodiacetic acid, L × I.D. 15 cm × 21.5 mm, 13 μm particle size

material

stainless steel

product line

TSKgel®

form

slurry

manufacturer/tradename

Tosoh 8646

extent of labeling

20 μmol/mL resin ligand density

parameter

≤20% organic solvent
≤3 M salt concentration
≤45 °C temperature
1.0 mL/min flow rate
15 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 21.5 mm

matrix

hydroxylated methacrylic polymer

matrix active group

iminodiacetic acid phase

particle size

13 μm

operating pH

2-12

separation technique

affinity

Looking for similar products? Visit Product Comparison Guide

General description

High efficiency, resin-based TSKgel affinity columns (10 μm particles) separate or purify many enzymes and other proteins.

TSKgel Chelate-5PW columns utilize the tendency of iminodiacetic acid (IDA) to form a complex with metal ions, such as Zn2+, Ni2+ and Cu2+. The column is loaded with divalent metal ions by chelation. Peptides and proteins containing histidine residues will normally adsorb to these chelated ions at neutral pH. The retained compounds can be eluted with buffer containing imidazole or glycine. The key to making successful use of this retention mechanism is the proper selection of metal ions for chelation and the elution buffer to desorb the analytes. In general, Cu2+, interacts better with proteins; however, resolution is usually enhanced with Zn2+. A gradient mobile phase containing increasing imidazole or glycine concentrations is used to elute the retained compounds. Alternatively, a decreasing pH gradient can be used for elution purposes as well.

Applications include: immunoglobulins, transferrin, lectins, milk proteins, membrane proteins and peptides.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service