Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

807471

TOYOPEARL® HW-75S Bulk Media

phase hydroxylated methacrylic polymer, bottle of 250 mL, 30 μm particle size

Synonym(s):

TOYOPEARL® HW-75 Size Exclusion Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
Product 807471 is not currently sold in your country. Contact Technical Service

product line

TOYOPEARL®

form

slurry

packaging

bottle of 250 mL

manufacturer/tradename

Tosoh 7471

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

hydroxylated methacrylic polymer phase

particle size

30 μm

operating pH

2-13

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl HW-75 resin is a hydroxylated methacrylic polymer bead for size exclusion chromatography of proteins between 500,000-50,000,000 Da.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service