Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

807134

TSKgel® ODS-120T (10 µm) HPLC Columns

L x I.D. 30 cm × 21.5 mm, C18 (octadecyl) phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$4,960.00

$4,960.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 EA
$4,960.00

About This Item

UNSPSC Code:
41115711

$4,960.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

Product Name

TSKgel® ODS-120T HPLC Column, phase C18 (octadecyl), L × I.D. 30 cm × 21.5 mm, 10 μm particle size

material

stainless steel

agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh 07134

extent of labeling

22% Carbon, multilayer, encapped

parameter

0-100% organic solvent
10-50 °C temperature
12 mL/min flow rate
60 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

30 cm × 21.5 mm

matrix

spherical silica

matrix active group

C18 (octadecyl) phase

particle size

10 μm

pore size

150 Å

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel® ODS-120T columns contain 150Å pore size silica particles. The exclusion limit of 10,000 Da makes TSKgel ODS-120T columns a good choice for the reversed phase chromatography of peptides, small proteins, and small molecular weight compounds in organic and environmental samples. The columns are endcapped with trimethyl silane groups to improve the peak shape of positively charged analytes.

Physical form

Shipped in 70% methanol - 30% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service