Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78076

Supelco

Amberlite HPR4200 Ion Exchange Resin

chloride form, 25-30 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$47.60

$47.60


Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$47.60

About This Item

UNSPSC Code:
23151817

$47.60


Available to ship onApril 29, 2025Details


Request a Bulk Order

Product Name

Amberlite HPR4200, chloride form

form

beads

technique(s)

LPLC: suitable

loss

49.0-55.0% loss on drying

matrix

PS-DVB

matrix active group

SAX

particle size

0.600-0.700 mesh

capacity

≥1.30 meq/mL exchange capacity

separation technique

anion exchange

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service