Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73079

Supelco

11-Dodecenoic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
MDL number:

grade

analytical standard

assay

≥98.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

OC(CCCCCCCCCC=C)=O

InChI

1S/C12H22O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2H,1,3-11H2,(H,13,14)

InChI key

GZZPOFFXKUVNSW-UHFFFAOYSA-N

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service