Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73034AST

Supelco

Astec® CHIRALDEX G-TA Capillary GC Column

L × I.D. 40 m × 0.25 mm, df 0.12 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
eCl@ss:
32110502
NACRES:
SB.54

material

fused silica

description

GC capillary column

packaging

pkg of 1 ea

parameter

-10-180 °C temperature (isothermal or programmed)

Beta value

500

df

0.12 μm

technique(s)

gas chromatography (GC): suitable

L × I.D.

40 m × 0.25 mm

matrix active group

non-bonded; 2,6-di-O-pentyl-3-trifluoroacetyl derivative of γ-cyclodextrin phase

application(s)

agriculture
chemicals and industrial polymers
cleaning products
clinical
cosmetics
environmental
flavors and fragrances
food and beverages
forensics and toxicology
life science and biopharma
personal care
pharmaceutical (small molecule)

column type

capillary chiral

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

Astec® CHIRALDEX G-TA is the first choice in the Group 1 CSPs (Surface Interactions, Complex Derivatives). This phase has been shown to be the most broadly-selective phase for the pharmaceutical industry, especially for the analysis of chiral intermediates and drug studies in various stages of clinical trials. Separations occur without the inclusion mechanism and are typically faster and more efficient than most chiral stationary phases. G-TA has also been used to separate parent drug enantiomers and their metabolites. G-TA has its highest selectivity for oxygen-containing analytes like alcohols, diols and polyols as the free alcohol and as an acyl derivative; amines as acyl derivatives; amino alcohols, halogens (Cl>Br>F), amino acids, hydroxy acids, lactones, furans and pyrans. It is also highly selective for halogenated compounds.

Chem/Phys Resistance

Temp. Limits:
  • -10 °C to 180 °C isothermal and programmed

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic resolutions concentrate the minor enantiomer and aid measurement of high enantiomeric purity.
Caron, Gaetan; Tseng, George W.M.; Kazlauskas, R.J.
Tetrahedron Asymmetry, 5 (1), 83-92 (1994)
Asymmetric ring opening of meso-epoxides with B-halobis(2-isocaranyl)boranes 2-dIcr2BX
Roy, Chandra D., Brown, Herbert C.
Tetrahedron Asymmetry, 17 (13), 1931-1936 (2006)
Catalytic asymmetric synthesis of Japonilure and its enantiomer
Xu, Hao, et al.
Tetrahedron Asymmetry, 25 (20-21), 1372-1375 (2014)
Investigation of a novel diamine based chiral auxiliary in the asymmetric alkylation of ketones
Clarke, Sarah L, et al.
Tetrahedron Asymmetry, 25 (4), 356-361 (2014)
Hugo L van Beek et al.
FEBS open bio, 4, 168-174 (2014-03-22)
Enzyme stability is an important parameter in biocatalytic applications, and there is a strong need for efficient methods to generate robust enzymes. We investigated whether stabilizing disulfide bonds can be computationally designed based on a model structure. In our approach

Articles

Chromatographic enantiomeric separation of amino acids, like proline, is described for chiral GC analysis after derivatization.

Chromatograms

suitable for GC

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the regeneration procedure for Chiraldex TA columns?

    1 answer
    1. The regeneration procedure for ChiraldexTA columns is located in Supelco brochure, T407123 (JCH), A Guide to Using Cyclodextrin Bonded Phases for Chiral Separations by Capillary Gas Chromatography, on pages 8 and 9.  These pages are attached.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service