Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72672

Supelco

N-Isovalerylglycine

analytical standard

Synonym(s):

N-(3-Methyl-1-oxobutyl)glycine, N-(3-Methylbutanoyl)glycine, [(3-Methylbutanoyl)amino]acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO3
CAS Number:
Molecular Weight:
159.18
Beilstein/REAXYS Number:
1766622
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

assay

≥95% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

InChI

1S/C7H13NO3/c1-5(2)3-6(9)8-4-7(10)11/h5H,3-4H2,1-2H3,(H,8,9)(H,10,11)

InChI key

ZRQXMKMBBMNNQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Isovalerylglycine is a byproduct of the catabolism of the amino acid leucine. Accumulation of isovalerylglycine occurs in Isovaleric Acidemia (IVA), an autosomal recessive disorder caused by mutations in the isovaleryl-CoA dehydrogenase gene. The deficiency of this enzyme in the metabolism of leucine leads to the accumulation of a series of isovaleryl-CoA metabolites, such as isovalerylglycine (IVG).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service