Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72529

Supelco

Methyl 3-[[(2RS)-2-(dipropylamino)propanoyl]-amino]-4-methylthiophene-2-carboxylate hydrochloride

reference material

Synonym(s):

Articaine impurity H (PhEur), Dipropylarticaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26N2O3S · HCl
CAS Number:
Molecular Weight:
362.92
MDL number:
UNSPSC Code:
41116107

grade

reference material

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service