Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69638

Supelco

cis-2-Decenoic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
Beilstein/REAXYS Number:
1721990
MDL number:
UNSPSC Code:
12352211

grade

analytical standard

assay

≥98.0% (HPLC)

form

liquid

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.457-1.459

storage temp.

−20°C

SMILES string

CCCCCCC\C=C/C(O)=O

InChI

1S/C10H18O2/c1-2-3-4-5-6-7-8-9-10(11)12/h8-9H,2-7H2,1H3,(H,11,12)/b9-8-

InChI key

WXBXVVIUZANZAU-HJWRWDBZSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service