Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69573

Supelco

cis,cis-5,8-Tetradecadienoic acid

analytical standard

Synonym(s):

Goshuyic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24O2
CAS Number:
Molecular Weight:
224.34

grade

analytical standard

assay

≥94.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

OC(CCC/C=C\C/C=C\CCCCC)=O

InChI

1S/C14H24O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h6-7,9-10H,2-5,8,11-13H2,1H3,(H,15,16)/b7-6-,10-9-

InChI key

HXHZGHRLVRFQDR-HZJYTTRNSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service