Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65806-U

Supelco

Discovery® Zr-CarbonC18 Supelguard Cartridge

5 μm particle size, L × I.D. 1 cm × 2.1 mm, pkg of 2 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

material

stainless steel column

description

Supelguard Cartridge

product line

Discovery®

feature

endcapped: no

packaging

pkg of 2 ea

technique(s)

HPLC: suitable

L × I.D.

1 cm × 2.1 mm

surface area

30 m2/g

matrix

zirconia particle platform
polymeric

matrix active group

octadecylphenyl phase

particle size

5 μm

pore size

300 Å

operating pH

1-14

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service