Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65720-U

Supelco

Discovery® Zr-PBD HPLC Column

5 μm particle size, L × I.D. 15 cm × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

material

stainless steel column

agency

suitable for USP L49

product line

Discovery®

feature

endcapped: no

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

2% Carbon loading

parameter

≤100 °C temp. range (up to 150 °C with special hardware)
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 2.1 mm

matrix

zirconia, spherical, porous particle platform
polymeric

matrix active group

polybutadiene phase

particle size

5 μm

pore size

300 Å

operating pH range

1-13

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Discovery® Zr-PBD comprises spherical, porous zirconia particles with a durable coating of polybutadiene. It operates via a reverse-phase mechanism, but is less hydrophobic, so less organic solvent is required for elution. Discovery Zr-PBD complements the selectivity offering of the other zirconia and silica-based Discovery phases, and allows the use of a wide range of mobile phase pH from pH 1 to 13.

Features and Benefits

• General purpose zirconia phase
• Selectivity similar to C18-silica

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service