Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65712-U

Supelco

Discovery® Zr-SAX HPLC Column

3 μm particle size, L × I.D. 10 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

material

stainless steel column

product line

Discovery®

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

25% Carbon loading

parameter

<80 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

10 cm × 4.6 mm

surface area

30 m2/g

matrix

polymeric
zirconia spherical, porous particle platform

matrix active group

polyethyleneimine bonding phase

particle size

3 μm

pore size

300 Å

operating pH range

1-12

application(s)

food and beverages

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Efficient strong anion-exchanger useful for inorganic and organic anions. Stable from pH 1-12.
  • Ideal for the separation of water-soluble vitamins.
  • Useful for the separation of bio-molecules such as nucleotides, nucleosides, oligonucleotides, oligodeonucleotides, amino acids and peptides.
  • High anion-exchange capacity that can be controlled by the amount of polymer deposited on the porous zirconia substrate.
  • Does not shrink or swell as a function of anionic strength or organic modifier content of the mobile phase.
  • Extremely stable amino phase for normal phase separation of carbohydrates.
  • Thermally stable up to 80°C, which causes different selectivity and high speed separations with lower ionic strength mobile phases. This is very important in the preparation of RNA and DNA samples used for further studies.
  • Mixed-mode separation modes may be exploited to optimize separations, including Lewis acid-base interactions, hydrophobic interactions and ion-exchange interactions. These modes may be attenuated by adjusting the strong Lewis base content, organic content and ionic strength of the mobile phase, respectively.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service