Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64804-U

Supelco

Separatory Funnel

glass stopcock plug

, size 2000 mL

Synonym(s):

SEPARATORY FUNNEL 2000ML W/GLASS STOPCOCK, EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NB.21

size

38 (stopper)
6 (stopcock)

size

2000 mL

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

Squibb-type, with glass or PTFE stopcock plug.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service