Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64699-U

Supelco

Micro Connector

for connecting 13mm to 13mm threaded joint

Synonym(s):

Micro Connector, 13mm to 13mm, pk 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NB.21

packaging

pkg of 6 ea

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for connecting 13mm to 13mm threaded joint
for use with Kuderna-Danish Sample Concentrator

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service