Key Documents
64688-U
Supelco
Kuderna-Danish Sample Concentrator
Micro design, threaded joint
Synonym(s):
KIT-MICRO-CONCENTRATOR
About This Item
UNSPSC Code:
41115724
NACRES:
NB.21
General description
Widely used in semivolatile and pesticide analysis
For US EPA protocol
Developed for concentrating compounds dissolved in
volatile solvents
Made in our own glass shop
Choose from one of the most recent versions:
Certificates of Analysis (COA)