Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64678-U

Supelco

Round Bottom Flask

size 500 mL

Synonym(s):

ROUND BOTTOM FLASK 500ML WITH 24/40 GR.JT.,EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115724
eCl@ss:
32060304
NACRES:
NB.21

material

round bottom glass flask

joint

joint ST/NS flask

packaging

pkg of 1 ct

size

500 mL

compatibility

for use with Liquid/Liquid Extractor Kit (64796-U)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service