Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59601-U

Supelco

Discovery® BIO PolyMA-SCX Column

5 μm particle size, L × I.D. 5 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

manufacturer/tradename

Discovery®

parameter

4-50 °C temp. range
735 psi max. pressure

technique(s)

HPLC: suitable

L × I.D.

5 cm × 4.6 mm

surface coverage

surface coverage 0.3 meq/g

matrix

polymethacrylate, spherical, monodispersed particle platform

matrix active group

sulfopropyl phase, Na+ counter ion

particle size

5 μm

pore size

1000 Å

operating pH range

1-13

compatibility

mode of use cation-exchange

separation technique

cation exchange

Application

For use at pH less than the protein isoelectric point (pI), usually at pH less than 7

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service