Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59456

Opti-Max® outlet check valve cartridge system

for use with 1050,1100, Agilent/HP 01018-60008, pkg of 1 ea

Synonym(s):

Optimize Technologies HPLC Pump Part for Agilent/HP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

material

stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

Agilent/HP 01018-60008

compatibility

for use with 1050,1100

Looking for similar products? Visit Product Comparison Guide

Legal Information

Opti-Max is a registered trademark of Optimize Technologies, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service