Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59408

Sapphire plunger

for use with 1050,1100, pkg of 1 ea, Agilent/HP 55062-2441

Synonym(s):

Optimize Technologies HPLC Pump Part for Agilent/HP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

packaging

pkg of 1 ea

manufacturer/tradename

Agilent/HP 55062-2441

compatibility

for use with 1050,1100

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service