Key Documents
59284
SSI Flow-Through Back-Pressure Regulator
About This Item
UNSPSC Code:
41115700
General description
Designed to apply constant pressure to an HPLC detector outlet, over a wide range of mobile phase flows and viscosities. Can be used with flowmeters. Minimizes bubble formation in the detector cell, improves baseline stability. Flow-through design minimizes band spreading. Immune to clogging. Adjustable from 5 to 75 psi, for compatibility with any detector. The flow through design has wetted parts constructed of a solvent resistant, non metallic, biocompatible material.
Legal Information
SSI is a trademark of Scientific Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
András Domokos et al.
International journal of pharmaceutics, 581, 119297-119297 (2020-04-04)
An end-to-end continuous pharmaceutical manufacturing process was developed for the production of conventional direct compressed tablets on a proof-of-concept level for the first time. The output reaction mixture of the flow synthesis of acetylsalicylic acid was crystallized continuously in a